Gene Transcripts

Gene Symbol
ARL14
Entrez Gene ID
80117
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
ARF7
Chromosome Location
Chr. 3: 160677160 - 160678448 on Build GRCh38
UniGene ID
Hs.287702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025047.2 NP_079323.1 1 269
GenBank mRNA AK026248.1 - 1 270
BC034354.1 - 1 252 4747382
BM760777.1 - 1 270

View Full Product Details