Gene Transcripts
- Gene Symbol
- TMEM254
- Entrez Gene ID
- 80195
- Gene Name
- transmembrane protein 254
- Gene Aliases
- C10orf57, bA369J21.6
- Chromosome Location
- Chr. 10: 80078646 - 80092552 on Build GRCh38
- UniGene ID
- Hs.169982
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270367.1 |
NP_001257296.1
|
3, 4 |
321 |
|
NM_001270371.1 |
NP_001257300.1
|
4, 5 |
395 |
|
NM_001270374.1 |
NP_001257303.1
|
4, 5 |
702 |
|
NM_025125.3 |
NP_079401.2
|
3, 4 |
300 |
|
NR_072984.1 |
|
4, 5 |
317 |
|
NR_072985.1 |
|
4, 5 |
324 |
|
NR_072986.1 |
|
3, 4 |
222 |
|
XM_006717978.2 |
XP_006718041.1
|
4, 5 |
652 |
|
GenBank mRNA |
AK222536.1 |
- |
3, 4 |
281 |
|
AK300490.1 |
- |
3, 4 |
331 |
|
AK309425.1 |
- |
4, 5 |
599 |
|
AK310160.1 |
- |
4, 5 |
282 |
|
AK311563.1 |
- |
4, 5 |
290 |
|
AL832397.1 |
- |
3, 4 |
197 |
|
AL832483.1 |
- |
3, 4 |
308 |
|
BC022252.1 |
- |
3, 4 |
276 |
4668550 |
BX537803.1 |
- |
4, 5 |
282 |
|
CR457310.1 |
- |
3, 4 |
246 |
|
DA880596.1 |
- |
4, 5 |
645 |
|
DB485367.1 |
- |
4, 5 |
324 |
|
View Full Product Details