Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025248.2 | NP_079524.2 | 19 | 3694 | |
XM_017025169.1 | XP_016880658.1 | 23 | 4786 | ||
XM_017025170.1 | XP_016880659.1 | 21 | 3984 | ||
XM_017025171.1 | XP_016880660.1 | 20 | 3861 | ||
XM_017025172.1 | XP_016880661.1 | 19 | 3759 | ||
GenBank mRNA | AB051471.1 | - | 13 | 2206 | |
AB127405.1 | - | 19 | 3694 |