Gene Transcripts

Gene Symbol
LCOR
Entrez Gene ID
84458
Gene Name
ligand dependent nuclear receptor corepressor
Gene Aliases
MLR2
Chromosome Location
Chr. 10: 96832250 - 96964441 on Build GRCh38
UniGene ID
Hs.745068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170765.1 NP_001164236.1 8 1617
NM_001170766.1 NP_001164237.1 8 1617
NM_032440.3 NP_115816.2 8 1637
XM_006718033.3 XP_006718096.1 8 1925
XM_017016784.1 XP_016872273.1 7 1548
XM_017016785.1 XP_016872274.1 9 1692
XM_017016786.1 XP_016872275.1 8 1613
XM_017016787.1 XP_016872276.1 7 1485
XM_017016788.1 XP_016872277.1 7 1539
GenBank mRNA AB058698.1 - 8 1550
AL832044.1 - 1 376
AL832106.1 - 8 1597
AL834245.2 - 8 1620
BC053359.1 - 8 1558 6009825

View Full Product Details