Gene Transcripts

Gene Symbol
LYZL1
Entrez Gene ID
84569
Gene Name
lysozyme like 1
Gene Aliases
KAAG648, LYC2, LYZD1, PRO1278, bA534G20.1
Chromosome Location
Chr. 10: 29289051 - 29318328 on Build GRCh38
UniGene ID
Hs.558572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032517.4 NP_115906.3 5 593
GenBank mRNA AY358694.1 - 5 538
BC021730.2 - 5 559 4825273

View Full Product Details