Gene Transcripts

Gene Symbol
GTPBP10
Entrez Gene ID
85865
Gene Name
GTP binding protein 10
Gene Aliases
ObgH2, UG0751c10
Chromosome Location
Chr. 7: 90346665 - 90391455 on Build GRCh38
UniGene ID
Hs.593547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042717.2 NP_001036182.1 1, 2 118
NM_033107.3 NP_149098.2 1, 2 118
GenBank mRNA AF351613.1 - 1, 2 119
AK294325.1 - 1, 2 68
AL525056.3 - 1, 2 60
BC004923.2 - 15
BC021573.1 - 15
BC144162.1 - 1, 2 65

View Full Product Details