Gene Transcripts
- Gene Symbol
- ATG16L2
- Entrez Gene ID
- 89849
- Gene Name
- autophagy related 16 like 2
- Gene Aliases
- ATG16B, WDR80
- Chromosome Location
- Chr. 11: 72814406 - 72843740 on Build GRCh38
- UniGene ID
- Hs.653186
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318766.1 |
NP_001305695.1
|
8, 9 |
1053 |
|
NM_033388.1 |
NP_203746.1
|
8, 9 |
933 |
|
XM_005274376.4 |
XP_005274433.1
|
8, 9 |
932 |
|
XM_006718733.3 |
XP_006718796.1
|
8, 9 |
932 |
|
XM_011545332.1 |
XP_011543634.1
|
7, 8 |
818 |
|
XM_011545333.1 |
XP_011543635.1
|
8, 9 |
1381 |
|
XM_011545334.1 |
XP_011543636.1
|
8, 9 |
794 |
|
GenBank mRNA |
AK090597.1 |
- |
8, 9 |
1044 |
|
AK091151.1 |
- |
7, 8 |
2521 |
|
AK093017.1 |
- |
8, 9 |
906 |
|
BC036713.2 |
- |
6, 7 |
662 |
|
BC137489.1 |
- |
8, 9 |
1031 |
9021489 |
BC137490.1 |
- |
8, 9 |
1031 |
9021490 |
View Full Product Details