Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
Chromosome Location
Chr. 4: 112539333 - 112636995 on Build GRCh38
UniGene ID
Hs.380346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018392.4 NP_060862.3 17 4695
XM_005263115.3 XP_005263172.1 17 4750
XM_011532091.2 XP_011530393.1 16 4644
XM_011532092.2 XP_011530394.1 16 4575
XM_011532093.2 XP_011530395.1 16 4521
XM_011532094.2 XP_011530396.1 15 4437
XM_011532096.2 XP_011530398.1 17 4695
XM_011532097.2 XP_011530399.1 10 1797
XM_011532098.2 XP_011530400.1 8 1471
XM_011532099.2 XP_011530401.1 7 1430
XM_017008369.1 XP_016863858.1 16 4690
XM_017008370.1 XP_016863859.1 15 4470
XM_017008371.1 XP_016863860.1 14 4350
XM_017008372.1 XP_016863861.1 14 3729
XM_017008373.1 XP_016863862.1 13 3666
GenBank mRNA AK126054.1 - 6 1079
AL137700.1 - 3 270
BX647594.1 - 6 1178

View Full Product Details