Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017919.1 | NP_001017919.1 | 5 | 962 | |
NM_033544.2 | NP_291022.2 | 6 | 1046 | ||
GenBank mRNA | BC064606.1 | - | 3 | 973 | |
BC094739.1 | - | 5 | 1249 | ||
BC113826.1 | - | 3 | 370 | ||
BC140708.1 | - | 5 | 960 | 9021576 | |
BC140709.1 | - | 5 | 960 | 9021577 | |
BU850147.1 | - | 4 | 763 |