Gene Transcripts

Gene Symbol
C12orf57
Entrez Gene ID
113246
Gene Name
chromosome 12 open reading frame 57
Gene Aliases
C10, GRCC10
Chromosome Location
Chr. 12: 6943438 - 6946003 on Build GRCh38
UniGene ID
Hs.405913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301834.1 NP_001288763.1 4 610
NM_001301836.1 NP_001288765.1 3 607
NM_001301837.1 NP_001288766.1 3 652
NM_001301838.1 NP_001288767.1 3 835
NM_138425.3 NP_612434.1 3 739
NR_126035.1 3 748
GenBank mRNA AV751928.1 - 490
BC009925.2 - 3 516 3537206
BE888740.1 - 3 526
BP318750.2 - 343

View Full Product Details