Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052886.2 | NP_443118.1 | 2 | 281 | |
GenBank mRNA | AK172820.1 | - | 2 | 197 | |
AK312030.1 | - | 2 | 188 | ||
AY007723.1 | - | 2 | 258 | ||
BC012367.1 | - | 2 | 281 | 3860009 | |
CV574204.1 | - | 2 | 281 |