Gene Transcripts

Gene Symbol
BTBD9
Entrez Gene ID
114781
Gene Name
BTB domain containing 9
Gene Aliases
dJ322I12.1
Chromosome Location
Chr. 6: 38168451 - 38640426 on Build GRCh38
UniGene ID
Hs.603858
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099272.1 NP_001092742.1 5 1344
NM_001172418.1 NP_001165889.1 4 1129
NM_052893.1 NP_443125.1 6 1544
NM_152733.2 NP_689946.2 4 1081
XM_011514279.2 XP_011512581.1 7 1941
XM_011514280.2 XP_011512582.1 5 1622
XM_011514281.2 XP_011512583.1 5 1622
XM_011514282.2 XP_011512584.1 5 1622
XM_017010227.1 XP_016865716.1 6 1822
GenBank mRNA AB067467.1 - 5 1210
AK057507.1 - 4 1081
AK090930.1 - 1 48
BC101354.2 - 4 1002 40025568
BC101355.1 - 4 1002 40025569
BC101356.2 - 4 1002
BC101357.1 - 4 1050 40025572
BX537534.1 - 5 1077

View Full Product Details