Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052947.3 | NP_443179.3 | 11 | 6274 | |
XM_011525801.2 | XP_011524103.1 | 11 | 6507 | ||
XM_011525802.2 | XP_011524104.1 | 9 | 6894 | ||
XM_017025535.1 | XP_016881024.1 | 11 | 6508 | ||
XM_017025536.1 | XP_016881025.1 | 11 | 6508 | ||
XM_017025537.1 | XP_016881026.1 | 10 | 6420 | ||
XM_017025538.1 | XP_016881027.1 | 8 | 5392 | ||
XM_017025539.1 | XP_016881028.1 | 8 | 4140 | ||
GenBank mRNA | AL832018.2 | - | 8 | 4606 | |
AY044450.1 | - | 8 | 4341 | ||
BX647446.1 | - | 7 | 3752 | ||
BX647639.1 | - | 8 | 5140 | ||
BX647796.1 | - | 11 | 6261 |