Gene Transcripts

Gene Symbol
MRPL53
Entrez Gene ID
116540
Gene Name
mitochondrial ribosomal protein L53
Gene Aliases
L53MT
Chromosome Location
Chr. 2: 74471958 - 74472815 on Build GRCh38
UniGene ID
Hs.534527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053050.4 NP_444278.1 2 306
GenBank mRNA BC012163.1 - 2 157
BG196300.1 - 2 291
BQ688110.1 - 2 163

View Full Product Details