Gene Transcripts

Gene Symbol
COLCA2
Entrez Gene ID
120376
Gene Name
colorectal cancer associated 2
Gene Aliases
C11orf93, CASC13, LOH11CR1G
Chromosome Location
Chr. 11: 111298517 - 111308735 on Build GRCh38
UniGene ID
Hs.31409
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136105.2 NP_001129577.1 4 759
NM_001271457.1 NP_001258386.1 4 504
NM_001271458.1 NP_001258387.1 4 328
XM_017017196.1 XP_016872685.1 5 408
XM_017017197.1 XP_016872686.1 3 1499
XM_017017198.1 XP_016872687.1 4 364
XM_017017199.1 XP_016872688.1 4 527
XM_017017200.1 XP_016872689.1 4 812
GenBank mRNA AK292195.1 - 4 759
BC042557.1 - 4 570
BC110079.1 - 3 227
BQ893717.1 - 534
HY114242.1 - 4 329
KF740510.1 - 4 704
KF740511.1 - 4 319
KF740512.1 - 5 424
KF740513.1 - 5 550
KF740514.1 - 5 902

View Full Product Details