Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001305121.1 | NP_001292050.1 | 4 | 1030 | |
NM_001305122.1 | NP_001292051.1 | 5 | 1095 | ||
NM_153208.2 | NP_694940.1 | 5 | 1095 | ||
NR_130967.1 | 5 | 1095 | |||
XM_011545736.2 | XP_011544038.1 | 4 | 430 | ||
XM_017022933.1 | XP_016878422.1 | 4 | 1141 | ||
GenBank mRNA | AF520569.1 | - | 5 | 1095 | |
AF520570.1 | - | 5 | 1095 | ||
AK000122.1 | - | 4 | 1030 | ||
AK093894.1 | - | 4 | 366 | ||
AK301933.1 | - | 4 | 418 | ||
AK303998.1 | - | 3 | 323 | ||
AK315672.1 | - | 4 | 418 | ||
BC034823.1 | - | 4 | 426 | 5165639 |