Gene Transcripts

Gene Symbol
SLC9B2
Entrez Gene ID
133308
Gene Name
solute carrier family 9 member B2
Gene Aliases
NHA2, NHE10, NHEDC2
Chromosome Location
Chr. 4: 103018033 - 103077371 on Build GRCh38
UniGene ID
Hs.546482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178833.5 NP_849155.2 4 934
XM_006714085.2 XP_006714148.1 4 933
GenBank mRNA AK172823.1 - 4 934
BC047447.1 - 4 464 5295722

View Full Product Details