Gene Transcripts

Gene Symbol
OR2F2
Entrez Gene ID
135948
Gene Name
olfactory receptor family 2 subfamily F member 2
Gene Aliases
OR7-1
Chromosome Location
Chr. 7: 143935233 - 143936186 on Build GRCh38
UniGene ID
Hs.553596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004685.1 NP_001004685.1 1 766
GenBank mRNA BC136812.1 - 1 833
BC136813.1 - 1 833 9020803

View Full Product Details