Gene Transcripts

Gene Symbol
FUNDC1
Entrez Gene ID
139341
Gene Name
FUN14 domain containing 1
Gene Aliases
Chromosome Location
Chr. X: 44523639 - 44542975 on Build GRCh38
UniGene ID
Hs.7549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173794.3 NP_776155.1 4, 5 539
GenBank mRNA BC035015.2 - 4, 5 423
BC042813.1 - 4, 5 534 5274638
BM837991.1 - 4, 5 565

View Full Product Details