Gene Transcripts

Gene Symbol
DTX3L
Entrez Gene ID
151636
Gene Name
deltex E3 ubiquitin ligase 3L
Gene Aliases
BBAP
Chromosome Location
Chr. 3: 122564338 - 122575203 on Build GRCh38
UniGene ID
Hs.518201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138287.3 NP_612144.1 3 1678
GenBank mRNA AF484416.1 - 3 1589
AK125086.1 - 3 1678
AL833598.1 - 3 1611
AY225123.1 - 3 1723
BC042191.1 - 3 1597 4339554
BC060509.1 - 3 1620 6457163
BX648267.1 - 1 1102
BX648645.1 - 3 1431

View Full Product Details