Gene Transcripts

Gene Symbol
SPICE1
Entrez Gene ID
152185
Gene Name
spindle and centriole associated protein 1
Gene Aliases
CCDC52, SPICE
Chromosome Location
Chr. 3: 113442718 - 113515187 on Build GRCh38
UniGene ID
Hs.477144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144718.3 NP_653319.1 10 1432
XM_011512461.2 XP_011510763.1 10 1189
XM_017005775.1 XP_016861264.1 10 1189
XM_017005776.1 XP_016861265.1 10 1326
XM_017005777.1 XP_016861266.1 10 1249
XM_017005778.1 XP_016861267.1 10 1433
GenBank mRNA AK126897.1 - 6 838
AL832148.1 - 10 1401
AL832327.2 - 10 1215
AY099107.1 - 10 1352
BC036951.1 - 10 1373 5500345

View Full Product Details