Gene Transcripts

Gene Symbol
TRAPPC3L
Entrez Gene ID
100128327
Gene Name
trafficking protein particle complex 3 like
Gene Aliases
BET3L, bA259P20.2
Chromosome Location
Chr. 6: 116496483 - 116545610 on Build GRCh38
UniGene ID
Hs.134795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139444.2 NP_001132916.1 2 212
GenBank mRNA BX280366.1 - 2 212
DY655209.1 - 1 19

View Full Product Details