Gene Transcripts
- Gene Symbol
- SFTPA2
- Entrez Gene ID
- 729238
- Gene Name
- surfactant protein A2
- Gene Aliases
- COLEC5, PSAP, PSP-A, PSPA, SFTP1, SFTPA2B, SP-2A, SP-A, SPA2, SPAII
- Chromosome Location
- Chr. 10: 79555852 - 79560461 on Build GRCh38
- UniGene ID
- Hs.523084
- Species
- Homo sapiens
- Gene Symbol
- SFTPA1
- Entrez Gene ID
- 653509
- Gene Name
- surfactant protein A1
- Gene Aliases
- COLEC4, PSAP, PSP-A, PSPA, SFTP1, SFTPA1B, SP-A, SP-A1, SPA, SPA1
- Chromosome Location
- Chr. 10: 79610939 - 79615443 on Build GRCh38
- UniGene ID
- Hs.535295
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC171875.1 |
- |
5 |
871 |
|
View Full Product Details