Gene Transcripts

Gene Symbol
SERPINB1
Entrez Gene ID
1992
Gene Name
serpin family B member 1
Gene Aliases
EI, ELANH2, HEL-S-27, HEL57, LEI, M/NEI, MNEI, PI-2, PI2
Chromosome Location
Chr. 6: 2832332 - 2842049 on Build GRCh38
UniGene ID
Hs.381167
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030666.3 NP_109591.1 7 1285
NR_073111.1 8 1417
NR_073112.1 7 1281
XM_011514333.1 XP_011512635.1 8 1508
XM_011514334.2 XP_011512636.1 7 1419
XM_011514335.2 XP_011512637.1 7 1274
GenBank mRNA AK223370.1 - 7 1098
AK291327.1 - 7 1098
AK298044.1 - 8 1227
AK304641.1 - 7 1130
AK312973.1 - 7 1096
BC009015.1 - 7 1083 3456154
BM785367.1 - 1 64
BT006928.1 - 6 1039
EU794624.1 - 6 1039
GQ472232.1 - 7 1083
M93056.1 - 7 1087

View Full Product Details