Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143804.1 | NP_001137276.1 | 3 | 605 | |
NM_178500.3 | NP_848595.1 | 3 | 478 | ||
XM_017024271.1 | XP_016879760.1 | 3 | 398 | ||
GenBank mRNA | AJ457189.1 | - | 3 | 502 | |
BC117187.1 | - | 2 | 145 | 40125738 | |
BC143884.1 | - | 2 | 272 |