Gene Transcripts
- Gene Symbol
- CFAP221
- Entrez Gene ID
- 200373
- Gene Name
- cilia and flagella associated protein 221
- Gene Aliases
- FAP221, PCDP1
- Chromosome Location
- Chr. 2: 119544392 - 119688610 on Build GRCh38
- UniGene ID
- Hs.496753
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271049.1 |
NP_001257978.1
|
15, 16 |
1639 |
|
NR_073132.1 |
|
16, 17 |
1784 |
|
NR_073133.1 |
|
14, 15 |
1538 |
|
XM_006712353.3 |
XP_006712416.1
|
15, 16 |
1679 |
|
XM_017003550.1 |
XP_016859039.1
|
15, 16 |
1679 |
|
XM_017003551.1 |
XP_016859040.1
|
15, 16 |
1806 |
|
XM_017003552.1 |
XP_016859041.1
|
15, 16 |
1692 |
|
XM_017003553.1 |
XP_016859042.1
|
15, 16 |
2025 |
|
XM_017003554.1 |
XP_016859043.1
|
14, 15 |
1521 |
|
XM_017003555.1 |
XP_016859044.1
|
14, 15 |
1521 |
|
XM_017003556.1 |
XP_016859045.1
|
14, 15 |
1524 |
|
XM_017003557.1 |
XP_016859046.1
|
13, 14 |
1454 |
|
XM_017003558.1 |
XP_016859047.1
|
15, 16 |
1679 |
|
XM_017003559.1 |
XP_016859048.1
|
12, 13 |
2650 |
|
GenBank mRNA |
AK126306.1 |
- |
2, 3 |
189 |
|
AK293665.1 |
- |
14, 15 |
1521 |
|
BC036530.1 |
- |
16, 17 |
1750 |
4828213 |
View Full Product Details