Gene Transcripts

Gene Symbol
CFAP221
Entrez Gene ID
200373
Gene Name
cilia and flagella associated protein 221
Gene Aliases
FAP221, PCDP1
Chromosome Location
Chr. 2: 119544392 - 119688610 on Build GRCh38
UniGene ID
Hs.496753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271049.1 NP_001257978.1 15 1574
NR_073132.1 16 1719
NR_073133.1 14 1473
XM_006712353.3 XP_006712416.1 15 1614
XM_017003550.1 XP_016859039.1 15 1614
XM_017003551.1 XP_016859040.1 15 1741
XM_017003552.1 XP_016859041.1 15 1627
XM_017003553.1 XP_016859042.1 15 1960
XM_017003554.1 XP_016859043.1 14 1456
XM_017003555.1 XP_016859044.1 14 1456
XM_017003556.1 XP_016859045.1 14 1459
XM_017003557.1 XP_016859046.1 13 1389
XM_017003558.1 XP_016859047.1 15 1614
XM_017003559.1 XP_016859048.1 12 2585
GenBank mRNA AK126306.1 - 2 124
AK293665.1 - 14 1456
BC036530.1 - 16 1685 4828213

View Full Product Details