Gene Transcripts

Gene Symbol
SPATA24
Entrez Gene ID
202051
Gene Name
spermatogenesis associated 24
Gene Aliases
CCDC161, T6441
Chromosome Location
Chr. 5: 139395269 - 139404204 on Build GRCh38
UniGene ID
Hs.373612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194296.1 NP_919272.1 1, 2 145
XM_005271916.4 XP_005271973.1 1, 2 262
XM_011543254.2 XP_011541556.1 1, 2 195
GenBank mRNA AK098740.1 - 1, 2 146
BC040037.1 - 1, 2 125
BC050563.1 - 1, 2 125 5751687
BM785501.1 - 1, 2 146

View Full Product Details