Gene Transcripts

Gene Symbol
OR5T2
Entrez Gene ID
219464
Gene Name
olfactory receptor family 5 subfamily T member 2
Gene Aliases
OR11-177
Chromosome Location
Chr. 11: 56232106 - 56233185 on Build GRCh38
UniGene ID
Hs.553627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004746.1 NP_001004746.1 1 739
GenBank mRNA BC136876.1 - 1 815

View Full Product Details