Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145306.2 | NP_660349.1 | 4 | 824 | |
XM_005269606.1 | XP_005269663.1 | 4 | 1108 | ||
XM_005269608.3 | XP_005269665.1 | 3 | 762 | ||
XM_011539455.2 | XP_011537757.1 | 4 | 1365 | ||
XM_017015836.1 | XP_016871325.1 | 5 | 996 | ||
GenBank mRNA | BC013587.1 | - | 4 | 813 |