Gene Transcripts
- Gene Symbol
- ATXN7L1
- Entrez Gene ID
- 222255
- Gene Name
- ataxin 7 like 1
- Gene Aliases
- ATXN7L4
- Chromosome Location
- Chr. 7: 105604774 - 105877232 on Build GRCh38
- UniGene ID
- Hs.745056
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020725.1 |
NP_065776.1
|
1, 2 |
209 |
|
NM_152749.3 |
NP_689962.1
|
1, 2 |
209 |
|
XM_005250220.3 |
XP_005250277.1
|
2, 3 |
292 |
|
XM_006715900.2 |
XP_006715963.1
|
2, 3 |
292 |
|
XM_006715901.2 |
XP_006715964.1
|
2, 3 |
292 |
|
XM_006715902.3 |
XP_006715965.1
|
2, 3 |
292 |
|
XM_006715903.3 |
XP_006715966.1
|
2, 3 |
292 |
|
XM_011515935.1 |
XP_011514237.1
|
2, 3 |
292 |
|
XM_011515937.2 |
XP_011514239.1
|
2, 3 |
292 |
|
XM_011515939.1 |
XP_011514241.1
|
2, 3 |
292 |
|
XM_011515940.1 |
XP_011514242.1
|
2, 3 |
292 |
|
XM_011515950.1 |
XP_011514252.1
|
2, 3 |
292 |
|
XM_011515954.2 |
XP_011514256.1
|
2, 3 |
292 |
|
XM_017011860.1 |
XP_016867349.1
|
1, 2 |
124 |
|
GenBank mRNA |
AB033044.1 |
- |
1, 2 |
222 |
|
AK095475.1 |
- |
1, 2 |
123 |
|
AK097574.1 |
- |
1, 2 |
122 |
|
BC030616.2 |
- |
1, 2 |
206 |
4796569 |
BQ431139.1 |
- |
1, 2 |
216 |
|
View Full Product Details