Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008779.1 | NP_001008779.1 | 1 | 68 | |
NM_001142634.1 | NP_001136106.1 | 2 | 182 | ||
NM_182756.3 | NP_877433.2 | 3 | 256 | ||
GenBank mRNA | AY820303.1 | - | 1 | 50 | |
AY820304.1 | - | 1 | 50 | ||
BC093005.1 | - | 2 | 182 | 4839330 |