Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135863.1 | NP_001129335.1 | 4 | 871 | |
NM_001135864.1 | NP_001129336.1 | 4 | 852 | ||
NM_173802.3 | NP_776163.1 | 4 | 932 | ||
GenBank mRNA | AK290248.1 | - | 4 | 853 | |
AL832339.1 | - | 4 | 858 | ||
BC039107.1 | - | 5 | 1323 | 4837913 | |
BC039535.1 | - | 4 | 932 | 5744381 |