Gene Transcripts

Gene Symbol
MIR9-3HG
Entrez Gene ID
254559
Gene Name
MIR9-3 host gene
Gene Aliases
LINC00925
Chromosome Location
Chr. 15: 89361579 - 89398487 on Build GRCh38
UniGene ID
Hs.136313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015411.1 2 193
NR_133001.1 3 884
GenBank mRNA AK125292.1 - 2 119
AL526389.3 - 3 884
DB020558.1 - 1 56
DB452761.1 - 1 19

View Full Product Details