Gene Transcripts

Gene Symbol
MDGA1
Entrez Gene ID
266727
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 1
Gene Aliases
GPIM, MAMDC3
Chromosome Location
Chr. 6: 37632508 - 37697990 on Build GRCh38
UniGene ID
Hs.437993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153487.3 NP_705691.1 3 1443
XM_006715056.3 XP_006715119.1 3 387
XM_017010734.1 XP_016866223.1 3 387
GenBank mRNA AF478693.1 - 3 1443
AK091149.1 - 2 80

View Full Product Details