Gene Transcripts

Gene Symbol
NBPF8
Entrez Gene ID
728841
Gene Name
neuroblastoma breakpoint family member 8
Gene Aliases
NBPF8P
Chromosome Location
Chr. 1: 120436351 - 120467844 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037501.2 NP_001032590.2 7 1178
NR_102404.1 7 1178
NR_102405.1 6 1075
GenBank mRNA AK307809.1 - 5 782
AY894561.1 - 3 367
AY894562.1 - 3 367
AY894563.1 - 2 241
AY894567.1 - 6 1075
AY894570.1 - 3 367
AY894572.1 - 7 1178
BC086308.1 - 7 1021 5453529

Gene Symbol
NBPF1
Entrez Gene ID
55672
Gene Name
neuroblastoma breakpoint family member 1
Gene Aliases
AB13, AB14, AB23, AD2, NBG, NBPF
Chromosome Location
Chr. 1: 16562427 - 16613605 on Build GRCh38
UniGene ID
Hs.467587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC094705.1 - 1 33

Gene Symbol
NBPF15
Entrez Gene ID
284565
Gene Name
neuroblastoma breakpoint family member 15
Gene Aliases
AB14, AG3, NBPF16
Chromosome Location
Chr. 1: 144421386 - 144459464 on Build GRCh38
UniGene ID
Hs.523572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170755.2 NP_001164226.1 9 1355
NM_173638.4 NP_775909.2 6 825
XM_017001077.1 XP_016856566.1 4 473
GenBank mRNA AK097180.1 - 9 1274
AK290302.1 - 3 284
AY894566.1 - 3 265
BC023087.1 - 6 682 3918932
CV024999.1 - 2 193
ES313907.1 - 3 384

Gene Symbol
NBPF26
Entrez Gene ID
101060684
Gene Name
neuroblastoma breakpoint family member 26
Gene Aliases
Chromosome Location
Chr. 1: 120805561 - 120842215 on Build GRCh38
UniGene ID
Hs.534675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011542529.2 XP_011540831.1 7 1036
XM_017003011.1 XP_016858500.1 7 1035
GenBank mRNA AK294944.1 - 1 19

View Full Product Details