Gene Transcripts

Gene Symbol
C3orf33
Entrez Gene ID
285315
Gene Name
chromosome 3 open reading frame 33
Gene Aliases
AC3-33
Chromosome Location
Chr. 3: 155762612 - 155806287 on Build GRCh38
UniGene ID
Hs.350846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308229.1 NP_001295158.1 3 288
NM_173657.2 NP_775928.1 4 415
XM_005247379.4 XP_005247436.1 3 1260
XM_011512710.2 XP_011511012.1 2 213
XM_011512711.2 XP_011511013.1 3 392
GenBank mRNA AF115515.1 - 3 251
AK055701.1 - 4 394
AK289890.1 - 3 288
AL832165.1 - 3 273
BC042198.1 - 3 250
BC064898.1 - 3 260 6008321
DA275323.1 - 3 288

View Full Product Details