Gene Transcripts

Gene Symbol
PRRT3
Entrez Gene ID
285368
Gene Name
proline rich transmembrane protein 3
Gene Aliases
Chromosome Location
Chr. 3: 9945542 - 9952415 on Build GRCh38
UniGene ID
Hs.598147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207351.4 NP_997234.3 4 2294
NR_134900.1 4 2294
XM_011533628.2 XP_011531930.1 4 2372
XM_011533629.2 XP_011531931.1 4 2352
XM_017006256.1 XP_016861745.1 3 4159
GenBank mRNA BC040508.1 - 4 2271
BC089447.1 - 4 2253 30563721

View Full Product Details