Gene Transcripts

Gene Symbol
SLFN14
Entrez Gene ID
342618
Gene Name
schlafen family member 14
Gene Aliases
BDPLT20
Chromosome Location
Chr. 17: 35545732 - 35560841 on Build GRCh38
UniGene ID
Hs.591193
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129820.1 NP_001123292.1 3 1478
XM_017024576.1 XP_016880065.1 5 1647
XM_017024577.1 XP_016880066.1 5 1669
XM_017024578.1 XP_016880067.1 4 1536
XM_017024579.1 XP_016880068.1 4 1569
GenBank mRNA BC140847.1 - 3 1475
BC157877.1 - 3 1479 9057094
BC157879.1 - 3 1486 9057095

View Full Product Details