Gene Transcripts

Gene Symbol
ARSH
Entrez Gene ID
347527
Gene Name
arylsulfatase family member H
Gene Aliases
sulfatase
Chromosome Location
Chr. X: 3006613 - 3033385 on Build GRCh38
UniGene ID
Hs.351533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011719.1 NP_001011719.1 1 77
GenBank mRNA AY875940.1 - 1 77

View Full Product Details