Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003561.2 | 9, 10 | 2419 | ||
NR_027768.1 | 6, 7 | 2565 | |||
GenBank mRNA | AK093485.1 | - | 1, 2 | 46 | |
AK131353.1 | - | 9, 10 | 2412 | ||
AL834175.1 | - | 6, 7 | 2318 | ||
BC150309.1 | - | 6, 7 | 2026 |