Gene Transcripts

Gene Symbol
TOPAZ1
Entrez Gene ID
375337
Gene Name
testis and ovary specific PAZ domain containing 1
Gene Aliases
C3orf77
Chromosome Location
Chr. 3: 44241886 - 44337071 on Build GRCh38
UniGene ID
Hs.132125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145030.1 NP_001138502.1 20 5103
XM_017006361.1 XP_016861850.1 18 4734
GenBank mRNA BX648094.1 - 19 3605

View Full Product Details