Gene Transcripts

Gene Symbol
NRROS
Entrez Gene ID
375387
Gene Name
negative regulator of reactive oxygen species
Gene Aliases
ELLP3030, GARPL1, LRRC33, UNQ3030
Chromosome Location
Chr. 3: 196639696 - 196662004 on Build GRCh38
UniGene ID
Hs.702186
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198565.2 NP_940967.1 3 770
GenBank mRNA AY358322.1 - 3 676
BC044233.1 - 3 681 5725567
BQ070837.1 - 3 758

View Full Product Details