Gene Transcripts

Gene Symbol
XKR5
Entrez Gene ID
389610
Gene Name
XK related 5
Gene Aliases
HARL2754, UNQ2754, XRG5A, XRG5BM
Chromosome Location
Chr. 8: 6808517 - 6835644 on Build GRCh38
UniGene ID
Hs.558840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289973.1 NP_001276902.1 5 904
NM_207411.4 NP_997294.3 4 766
XM_017013401.1 XP_016868890.1 3 1185
GenBank mRNA AY358489.1 - 4 637
AY534242.1 - 4 766
AY534243.1 - 5 758

View Full Product Details