Gene Transcripts

Gene Symbol
OR56A3
Entrez Gene ID
390083
Gene Name
olfactory receptor family 56 subfamily A member 3
Gene Aliases
OR56A2P, OR56A3P, OR56A6
Chromosome Location
Chr. 11: 5947347 - 5948294 on Build GRCh38
UniGene ID
Hs.553741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003443.2 NP_001003443.2 1 51

View Full Product Details