Gene Transcripts

Gene Symbol
OR6C75
Entrez Gene ID
390323
Gene Name
olfactory receptor family 6 subfamily C member 75
Gene Aliases
Chromosome Location
Chr. 12: 55365111 - 55366049 on Build GRCh38
UniGene ID
Hs.554554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005497.1 NP_001005497.1 1 631

View Full Product Details