Gene Transcripts

Gene Symbol
ALG11
Entrez Gene ID
440138
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1P, GT8
Chromosome Location
Chr. 13: 52012387 - 52029648 on Build GRCh38
UniGene ID
Hs.512963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004127.2 NP_001004127.2 2 172
GenBank mRNA AK025456.1 - 2 145
AK296747.1 - 2 162
BC111022.1 - 2 145
BC142998.1 - 2 171 8860455

View Full Product Details