Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011531855.2 | XP_011530157.1 | 1 | 28 | |
XM_011531857.2 | XP_011530159.1 | 1 | 26 | ||
XM_011531859.2 | XP_011530161.1 | 1 | 25 | ||
XM_011531860.2 | XP_011530162.1 | 1 | 25 | ||
XM_011531862.2 | XP_011530164.1 | 1 | 24 | ||
XM_011531863.2 | XP_011530165.1 | 1 | 21 | ||
XM_011531866.2 | XP_011530168.1 | 1 | 17 |