Gene Transcripts

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
Chromosome Location
Chr. 11: 111293389 - 111305048 on Build GRCh38
UniGene ID
Hs.729225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302644.1 NP_001289573.1 2 2609
NM_001302645.1 NP_001289574.1 2 2476
NM_001302646.1 NP_001289575.1 2 2647
NM_001302647.1 NP_001289576.1 2 2517
NM_001302648.1 NP_001289577.1 2 2482
NM_207429.3 NP_997312.1 2 2691
GenBank mRNA AB096245.1 - 2 2608
AK127703.1 - 1 334
CR936778.1 - 2 2606
KF728566.1 - 2 2609
KF728567.1 - 2 2476
KF728568.1 - 2 2691
KF728569.1 - 2 2609
KF728570.1 - 2 2647

View Full Product Details