Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001310338.1 | NP_001297267.1 | 6 | 1365 | |
NM_001310339.1 | NP_001297268.1 | 3 | 967 | ||
NM_052865.3 | NP_443097.1 | 5 | 1320 | ||
XM_005260867.2 | XP_005260924.1 | 6 | 1587 | ||
XM_005260870.3 | XP_005260927.1 | 4 | 1135 | ||
XM_006723663.3 | XP_006723726.1 | 5 | 1180 | ||
XM_011529394.1 | XP_011527696.1 | 6 | 1202 | ||
XM_011529395.1 | XP_011527697.1 | 5 | 1080 | ||
XM_017028127.1 | XP_016883616.1 | 5 | 1542 | ||
XM_017028128.1 | XP_016883617.1 | 5 | 1416 | ||
GenBank mRNA | AF306685.1 | - | 5 | 1129 | |
AK027503.1 | - | 5 | 1124 | ||
AK309563.1 | - | 3 | 663 | ||
AK309995.1 | - | 3 | 591 | ||
AK310282.1 | - | 4 | 636 | ||
AK315532.1 | - | 5 | 1156 | ||
BC016869.1 | - | 5 | 1092 | 3850853 | |
CV807140.1 | - | 3 | 780 |